EMAIL THIS PAGE TO A FRIEND
8.14518 EMD Millipore

Acetylferrocene

Acetylferrocene for synthesis. CAS 1271-55-2, molar mass 228.07 g/mol., for synthesis

  • CAS Number 1271-55-2

  • Linear Formula C₁₂H₁₂FeO

  • Molecular Weight 228.07 g/mol

  •  EC Number 215-043-2

Purchase

Properties

packaging   25 g in Glass bottle (8145180025)
bp   160 - 163 °C (4 hPa)
vapor pressure   <10 hPa (20 °C)
density   >1 g/cm3 (20 °C)
mp   82 - 85 °C
LD 50 oral   LD50 Rat 25 mg/kg
LD 50 dermal   LD50 Rat 50 - 500 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Melting range (lower value): ≥ 83 °C
  Melting range (upper value): ≤ 86 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2931 90 00
Hazard Statement(s) 
H300 + H310: Fatal if swallowed or in contact with skin
Precautionary Statement(s) 
P280: Wear protective gloves/ protective clothing.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
Storage class 
6.1A Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK 
WGK 3 highly water endangering
Disposal 
9
Carcinogenic compounds and flammable compounds labelled "Highly toxic" or "Toxic": container F. Alkyl sulfates are carcinogenic; Take particular care to avoid inhalation and skin contact. To neutralize alkyl sulfates, add dropwise (from a dropping funnel) to concentrated ice-cool Ammonia solution (Cat.No. 105426) with vigorous stirring. Before placing in container D, check the pH with pH Universal indicator strips (Cat.No. 109535).
Hazard Pictogram(s) 
GHS06  GHS06
Hazard Symbols 
Very toxic  Very toxic
Declaration (railroad and road) ADR, RID 
UN 2930 , 6.1 (4.1), II
Declaration (transport by air) IATA-DGR 
UN 2930 , 6.1 (4.1), II
Declaration (transport by sea) IMDG-Code 
UN 2930 , 6.1 (4.1), II
Categories of danger 
very toxic
R Phrase 
R 27/28
Very toxic in contact with skin and if swallowed.
S Phrase 
S 28-36/37-45
After contact with skin, wash immediately with plenty of water.Wear suitable protective clothing and gloves.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?