• USA Home
  • 8.14523 - 3,5-Dimethylpyrazol

EMAIL THIS PAGE TO A FRIEND
8.14523 EMD Millipore

3,5-Dimethylpyrazol

3,5-Dimethylpyrazol for synthesis. CAS 67-51-6, pH 6.7 (14 g/l, H₂O, 20 °C)., for synthesis

  • CAS Number 67-51-6

  • Linear Formula C₅H₈N₂

  • Molecular Weight 96.13 g/mol

  •  EC Number 200-657-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Plastic bottle (8145230025)
vapor pressure   <1 hPa (20 °C)
solubility   28.9 g/l soluble
density   1.14 g/cm3 (20 °C)
pH   6.7 (14 g/l, H₂O, 20 °C)
bulk density   340 kg/m3
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Melting range (lower value): ≥ 104 °C
  Melting range (upper value): ≤ 108 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 19 90
Hazard Statement(s) 
H302: Harmful if swallowed.
H373: May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement(s) 
P314: Get medical advice/ attention if you feel unwell.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Pictogram(s) 
GHS08  GHS08
Hazard Symbols 
Harmful  Harmful
R Phrase 
R 22-48/22
Harmful if swallowed.Harmful: danger of serious damage to health by prolonged exposure if swallowed.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?