• USA Home
  • 8.14526 - Benzyl acetoacetate

EMAIL THIS PAGE TO A FRIEND
8.14526 EMD Millipore

Benzyl acetoacetate

Benzyl acetoacetate for synthesis. CAS 5396-89-4, molar mass 192.21 g/mol., for synthesis

Synonym: Benzyl2-oxobutanoate

  • CAS Number 5396-89-4

  • Linear Formula C₁₁H₁₂O₃

  • Molecular Weight 192.21 g/mol

  •  EC Number 226-416-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 ml in Glass bottle (8145260100)
bp   276 °C
density   1.114 - 1.120 g/cm3 (20 °C)
flash point   108 °C
LD 50 oral   LD50 Rat 2600 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 95.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.114 - 1.120
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2918 30 00
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
S Phrase 
S 24/25
Avoid contact with skin and eyes.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?