• USA Home
  • 8.14540 - O-Methylhydroxylammonium chloride

EMAIL THIS PAGE TO A FRIEND
8.14540 EMD Millipore

O-Methylhydroxylammonium chloride

(25% solution in diluted HCl) for synthesis

Synonym: Methylhydroxylamine hydrochloride, Methoxyamine hydrochloride

Purchase

Properties

bp   105 - 110 °C (1013 hPa)
vapor pressure   117 hPa (50 °C)
density   1.12 g/cm3 (20 °C)
storage temp.   Store below +30°C.
specifications   Assay (acidimetric): 24.0 - 26.0 %
  Density (d 20 °C/ 4 °C): 1.100 - 1.110
  Free HCl (acidimetric): 17.0 - 19.0 %

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2928 00 90
Hazard Statement(s) 
H290: May be corrosive to metals.
H314: Causes severe skin burns and eye damage.
H317: May cause an allergic skin reaction.
H400: Very toxic to aquatic life.
Precautionary Statement(s) 
P273: Avoid release to the environment.
P280: Wear protective gloves/ protective clothing/ eye protection/ face protection.
P301 + P330 + P331: IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
Storage class 
8B Non-combustible, corrosive hazardous materials
WGK 
WGK 3 highly water endangering
Disposal 
28
Aqueous solutions: Container D.
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Pictogram(s) 
GHS09  GHS09
Hazard Symbols 
Corrosive  Corrosive
Hazard Symbols 
Dangerous for the environment  Dangerous for the environment
Declaration (railroad and road) ADR, RID 
UN 3265 , 8, II
Declaration (transport by air) IATA-DGR 
UN 3265 , 8, II
Declaration (transport by sea) IMDG-Code 
UN 3265 , 8, II, Segregation Group: 1 (Acids)
Categories of danger 
corrosive, sensitizing, dangerous for the environment
R Phrase 
R 34-43-50
Causes burns.May cause sensitisation by skin contact.Very toxic to aquatic organisms.
S Phrase 
S 26-36/37/39-45-61
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Wear suitable protective clothing, gloves and eye/face protection.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).Avoid release to the environment. Refer to special instructions/ Safety data sheets.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?