• USA Home
  • 8.14543 - Crown ether/4'-Nitrobenzo-15-crown-5

EMAIL THIS PAGE TO A FRIEND
8.14543 EMD Millipore

Crown ether/4'-Nitrobenzo-15-crown-5

Crown ether/4'-Nitrobenzo-15-crown-5 for synthesis. CAS 60835-69-0, molar mass 313.31 g/mol., for synthesis

  • CAS Number 60835-69-0

  • Linear Formula C₁₄H₁₉NO₇

  • Molecular Weight 313.31 g/mol

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   1 g in Glass bottle (8145430001)
mp   95 - 97 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Melting range (lower value): ≥ 94 °C
  Melting range (upper value): ≤ 97 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2932 99 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?