• USA Home
  • 8.14544 - 4,6,8-Trimethylazulene

EMAIL THIS PAGE TO A FRIEND
8.14544 EMD Millipore

4,6,8-Trimethylazulene

4,6,8-Trimethylazulene for synthesis. CAS 941-81-1, molar mass 170.25 g/mol., for synthesis

  • CAS Number 941-81-1

  • Linear Formula C₁₃H₁₄

  • Molecular Weight 170.25 g/mol

  •  EC Number 213-383-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   1 g in Glass bottle (8145440001)
mp   80 - 82 °C
shipped in   ambient
storage temp.   Store at +2°C to +8°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Melting range (lower value): ≥ 80 °C
  Melting range (upper value): ≤ 82 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2902 19 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Declaration (railroad and road) ADR, RID 
Kein Gefahrgut
Declaration (transport by air) IATA-DGR 
No Dangerous Good
Declaration (transport by sea) IMDG-Code 
No Dangerous Good
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?