• USA Home
  • 8.14556 - 2,3,4,5,6-Pentafluorobenzylalcohol

EMAIL THIS PAGE TO A FRIEND
8.14556 EMD Millipore

2,3,4,5,6-Pentafluorobenzylalcohol

2,3,4,5,6-Pentafluorobenzylalcohol for synthesis. CAS 440-60-8, molar mass 198.09 g/mol., for synthesis

  • CAS Number 440-60-8

  • Linear Formula C₇H₃F₅O

  • Molecular Weight 198.09 g/mol

  •  EC Number 207-126-7

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 g in Glass bottle (8145560005)
bp   114 - 115 °C (80 hPa)
flash point   86 °C
mp   30 - 33 °C
LD 50 oral   LD50 Rat 900 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Melting range (lower value): ≥ 30 °C
  Melting range (upper value): ≤ 33 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2906 29 00
Hazard Statement(s) 
H302: Harmful if swallowed.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Categories of danger 
harmful
R Phrase 
R 22
Harmful if swallowed.
S Phrase 
S 24/25
Avoid contact with skin and eyes.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?