• USA Home
  • 8.14557 - (S)-(-)-Butyllactate

EMAIL THIS PAGE TO A FRIEND
8.14557 EMD Millipore

(S)-(-)-Butyllactate

(S)-(-)-Butyllactate for synthesis. CAS 34451-19-9, molar mass 146.18 g/mol., for synthesis

  • CAS Number 34451-19-9

  • Linear Formula C₇H₁₄O₃

  • Molecular Weight 146.18 g/mol

  •  EC Number 252-036-3

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 ml in Glass bottle (8145570101)
bp   187 - 189 °C (1013 hPa)
vapor pressure   1 hPa (20 °C)
solubility   34 g/l
density   0.98 g/cm3 (20 °C)
flash point   79 °C
autoignition temp.   300 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.981 - 0.983
  Spec. rotation [α²0/D (undiluted): -13.5 - -11.0 °
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2918 11 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
S Phrase 
S 24/25
Avoid contact with skin and eyes.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?