EMAIL THIS PAGE TO A FRIEND
8.14558 EMD Millipore

4-Nitrophenol

4-Nitrophenol (stabilized with max. 5% water) for synthesis. CAS 100-02-7, chemical formula O₂NC₆H₄OH., (stabilized with max. 5% water) for synthesis

  • CAS Number 100-02-7

  • Linear Formula C₆H₅NO₃

  • Molecular Weight 139.11 g/mol

  •  EC Number 202-811-7

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 g in Glass bottle (8145580100)
bp   279 °C (1013 hPa) (decomposition),(anhydrous substance)
solubility   14.8 g/l (anhydrous substance)
density   1.48 g/cm3 (20 °C) (anhydrous substance)
flash point   169 °C (anhydrous substance)
autoignition temp.   510 °C (anhydrous substance)
mp   110 - 115 °C (anhydrous substance)
pH   4.4 (5 g/l, H₂O, 24 °C) (anhydrous substance)
LD 50 oral   LD50 Rat 202 mg/kg
LD 50 dermal   LD50 Rat 1024 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Melting point (after drying): 110 - 115 °C
  Water (K. F.): ≤ 5.0 %
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2908 99 00
Hazard Statement(s) 
H302 + H312 + H332: Harmful if swallowed, in contact with skin or if inhaled
H373: May cause damage to organs through prolonged or repeated exposure if swallowed.
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P314: Get medical advice/ attention if you feel unwell.
RTECS 
SM2275000
Storage class 
6.1C Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Pictogram(s) 
GHS08  GHS08
Hazard Symbols 
Harmful  Harmful
Declaration (railroad and road) ADR, RID 
UN 1663 , 6.1, III
Declaration (transport by air) IATA-DGR 
UN 1663 , 6.1, III
Declaration (transport by sea) IMDG-Code 
UN 1663 , 6.1, III
Categories of danger 
harmful
R Phrase 
R 20/21/22-33
Harmful by inhalation, in contact with skin and if swallowed.Danger of cumulative effects.
S Phrase 
S 28
After contact with skin, wash immediately with plenty of soap and water.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?