• USA Home
  • 8.14559 - 2-Phenylbenzothiazole

EMAIL THIS PAGE TO A FRIEND
8.14559 EMD Millipore

2-Phenylbenzothiazole

2-Phenylbenzothiazole for synthesis. CAS 883-93-2, molar mass 211.28 g/mol., for synthesis

  • CAS Number 883-93-2

  • Linear Formula C₁₃H₉NS

  • Molecular Weight 211.28 g/mol

  •  EC Number 212-935-3

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 g in Glass bottle (8145590005)
mp   112 - 115 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 99.0 % (a/a)
  Melting range (lower value): ≥ 112 °C
  Melting range (upper value): ≤ 115 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2934 20 80
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?