• USA Home
  • 8.14560 - Ethyl bromopyruvate

EMAIL THIS PAGE TO A FRIEND
8.14560 EMD Millipore

Ethyl bromopyruvate

Ethyl bromopyruvate for synthesis. CAS 70-23-5, molar mass 195.01 g/mol., for synthesis

  • CAS Number 70-23-5

  • Linear Formula C₅H₇BrO₃

  • Molecular Weight 195.01 g/mol

  •  EC Number 200-729-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8145600025)
bp   98 - 100 °C (13 hPa)
density   1.55 g/cm3 (20 °C)
flash point   99 °C
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 70.0 % (a/a)
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2918 30 00
Hazard Statement(s) 
H315: Causes skin irritation.
H319: Causes serious eye irritation.
H335: May cause respiratory irritation.
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P304 + P340: IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Categories of danger 
irritant
R Phrase 
R 36/37/38
Irritating to eyes, respiratory system and skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?