• USA Home
  • 8.14563 - Petroleum benzine boiling range 140-180°C

EMAIL THIS PAGE TO A FRIEND
8.14563 EMD Millipore

Petroleum benzine boiling range 140-180°C

Petroleum benzine boiling range 140-180°C EMPLURA®. CAS 64742-82-1., EMPLURA®

Synonym: Petroleum spirit, Petroleum ether

Purchase

Properties

Related Categories ReagentPlus and EMPLURA, Carbon Steel Flex-Spout Cans, Semi-Bulk Solvents, Solvent Packaging Options, Solvents More...
packaging   1 l in Glass bottle (8145631000)
bp   140 - 180 °C (1013 hPa)
expl. lim.     0.7 - 6.5 %(V)
vapor pressure   8 hPa (20 °C)
solubility   0.04 g/l
density   0.77 g/cm3 (20 °C)
flash point   24 °C
autoignition temp.   240 °C
mp   <-15 °C
shipped in   ambient
storage temp.   Store at +5°C to +30°C.
specifications   Density (d 20 °C/ 4 °C): 0.765 - 0.775
  Boiling range: 140 - 180 °C
  Identity (IR): passes test

Description

Other Notes

EMPLURA® is our low-cost alternative to high-purity qualities. With EMPLURA® we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA® solvents provide adequate specifications with the most important parameters.

Price and Availability

Safety & Documentation

Safety Information

HS Code 
2710 12 21
Hazard Statement(s) 
H226: Flammable liquid and vapour.
H304: May be fatal if swallowed and enters airways.
H372: Causes damage to organs through prolonged or repeated exposure.
Precautionary Statement(s) 
P210: Keep away from heat.
P301 + P330 + P331: IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
P314: Get medical advice/ attention if you feel unwell.
RTECS 
QK7945000
Storage class 
3 Flammable liquids
WGK 
WGK 1 slightly water endangering
Disposal 
1
Strongly contaminated halogen-free organic solvents: container A.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Pictogram(s) 
GHS08  GHS08
Hazard Symbols 
Harmful  Harmful
Declaration (railroad and road) ADR, RID 
UN 1268 , 3, III
Declaration (transport by air) IATA-DGR 
UN 1268 , 3, III
Declaration (transport by sea) IMDG-Code 
UN 1268 , 3, III
Categories of danger 
flammable, harmful
R Phrase 
R 10-48/20-65
Flammable.Harmful: danger of serious damage to health by prolonged exposure through inhalation.Harmful: may cause lung damage if swallowed.
S Phrase 
S 24-62-23
Avoid contact with skin.If swallowed, do not induce vomiting: seek medical advice immediately and show this container or label.Do not breathe vapour.
Protocols & Articles

Articles

Solvents for Classical Analysis, Production and Purification

Just the Right Solvent for Your Application For each of your applications, you want to use solvents that perfectly fit your needs. That’s why we strive to tailor our comprehensive range of solvents t...
Keywords: Chromatography, Evaporation, Organic synthesis, Solvents

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?