• USA Home
  • 8.14568 - 1,5-Hexadiene-3,4-diol

EMAIL THIS PAGE TO A FRIEND
8.14568 EMD Millipore

1,5-Hexadiene-3,4-diol

1,5-Hexadiene-3,4-diol (mixture of DL- and meso form) (stabilised with Hydroquinone) for synthesis. CAS 1069-23-4, molar mass 114.14 g/mol., (mixture of DL- and meso form) (stabilised with Hydroquinone) for synthesis

  • CAS Number 1069-23-4

  • Linear Formula C₆H₁₀O₂

  • Molecular Weight 114.14 g/mol

  •  EC Number 213-955-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 ml in Glass bottle (8145680005)
bp   198 °C (1013 hPa)
density   1.02 g/cm3 (20 °C)
flash point   93 °C
mp   14 °C
LD 50 oral   LD50 Rat 1652 mg/kg
LD 50 dermal   LD50 Rabbit 408 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay isomers (GC, area%): ≥ 96.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.017 - 1.020
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2905 39 95
Hazard Statement(s) 
H302: Harmful if swallowed.
H311: Toxic in contact with skin.
Precautionary Statement(s) 
P280: Wear protective gloves/ protective clothing.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 2 water endangering
Disposal 
9
Carcinogenic compounds and flammable compounds labelled "Highly toxic" or "Toxic": container F. Alkyl sulfates are carcinogenic; Take particular care to avoid inhalation and skin contact. To neutralize alkyl sulfates, add dropwise (from a dropping funnel) to concentrated ice-cool Ammonia solution (Cat.No. 105426) with vigorous stirring. Before placing in container D, check the pH with pH Universal indicator strips (Cat.No. 109535).
Hazard Pictogram(s) 
GHS06  GHS06
Hazard Symbols 
Harmful  Harmful
Declaration (railroad and road) ADR, RID 
UN 2810 , 6.1, III
Declaration (transport by air) IATA-DGR 
UN 2810 , 6.1, III
Declaration (transport by sea) IMDG-Code 
UN 2810 , 6.1, III
Categories of danger 
harmful
R Phrase 
R 21/22
Harmful in contact with skin and if swallowed.
S Phrase 
S 36/37
Wear suitable protective clothing and gloves.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?