• USA Home
  • 8.14569 - 2,5-Dimethylpyrazine

EMAIL THIS PAGE TO A FRIEND
8.14569 EMD Millipore

2,5-Dimethylpyrazine

2,5-Dimethylpyrazine for synthesis. CAS 123-32-0, pH 7 (H₂O)., for synthesis

Synonym: 2,5-Dimethyl paradiazine

  • CAS Number 123-32-0

  • Linear Formula C₆H₈N₂

  • Molecular Weight 108.14 g/mol

  •  EC Number 204-618-3

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8145690025)
  5 ml in Glass bottle (8145690005)
vapor pressure   4 hPa (20 °C)
density   0.99 g/cm3 (20 °C)
flash point   64 °C
mp   15 °C
pH   7 (H₂O)
LD 50 oral   LD50 Rat 1020 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.987 - 0.989
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 99 80
Hazard Statement(s) 
H302: Harmful if swallowed.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Categories of danger 
harmful
R Phrase 
R 22
Harmful if swallowed.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?