• USA Home
  • 8.14572 - 4-Acetamidobenzoic acid

EMAIL THIS PAGE TO A FRIEND
8.14572 EMD Millipore

4-Acetamidobenzoic acid

4-Acetamidobenzoic acid for synthesis. CAS 556-08-1, molar mass 179.17 g/mol., for synthesis

  • CAS Number 556-08-1

  • Linear Formula C₉H₉NO₃

  • Molecular Weight 179.17 g/mol

  •  EC Number 209-114-7

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Plastic bottle (8145720025)
mp   259 - 262 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 99.0 % (a/a)
  Melting range (lower value): ≥ 259 °C
  Melting range (upper value): ≤ 262 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2924 29 70
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
4
Liquid organic acids or solutions should be diluted, if necessary, and carefully neutralized with sodium hydrogen carbonate (Cat. No. 106323) or with sodium hydroxide (Cat. No 106462). Before filling into container D, check the pH with pH universal indicator strips (Cat. No. 109535).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?