• USA Home
  • 8.14615 - (R)-(-)-2-Amino-3-methylbutanol

EMAIL THIS PAGE TO A FRIEND
8.14615 EMD Millipore

(R)-(-)-2-Amino-3-methylbutanol

for synthesis

Synonym: D-Valinol

  • CAS Number 4276-09-9

  • Linear Formula C₅H₁₃NO

  • Molecular Weight 103.16 g/mol

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   1 g in Glass bottle (8146150001)
flash point   90 °C
mp   30 - 34 °C
shipped in   ambient
storage temp.   Store at +2°C to +8°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Melting range (lower value): ≥ 30 °C
  Melting range (upper value): ≤ 34 °C
  Spec. rotation [α²0/D (c=10 in water): -12.0 - -10.0 °
  Identity (IR): passes test
  Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2922 19 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Declaration (railroad and road) ADR, RID 
Kein Gefahrgut
Declaration (transport by air) IATA-DGR 
No Dangerous Good
Declaration (transport by sea) IMDG-Code 
No Dangerous Good
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?