• USA Home
  • 8.14616 - Dimethyl ammonium dimethylcarbamate

EMAIL THIS PAGE TO A FRIEND
8.14616 EMD Millipore

Dimethyl ammonium dimethylcarbamate

for synthesis

Synonym: Dimethylamine/carbon dioxide complex, N,N-Dimethylcarbamido acid dimethyl ammonium salt

  • CAS Number 4137-10-4

  • Linear Formula C₅H₁₄N₂O₂

  • Molecular Weight 134.18 g/mol

Purchase

Properties

Related Categories Biochemicals and Reagents More...
bp   60 - 61 °C
density   1.05 g/cm3 (20 °C)
flash point   13 °C DIN 51758
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.045 - 1.055
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2942 00 00
Hazard Statement(s) 
H225: Highly flammable liquid and vapour.
H315: Causes skin irritation.
H318: Causes serious eye damage.
H332: Harmful if inhaled.
H335: May cause respiratory irritation.
Precautionary Statement(s) 
P210: Keep away from heat/sparks/open flames/hot surfaces. No smoking.
P280: Wear eye protection.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P304 + P340: IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P313: Get medical advice/ attention.
P403 + P235: Store in a well-ventilated place. Keep cool.
Storage class 
3 Flammable liquids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Flammable  Flammable
Hazard Symbols 
Harmful  Harmful
Declaration (railroad and road) ADR, RID 
UN 2733 , 3 (8), II
Declaration (transport by air) IATA-DGR 
UN 2733 , 3 (8), II
Declaration (transport by sea) IMDG-Code 
UN 2733 , 3 (8), II, Segregation Group: 18 (Alkalis)
Categories of danger 
highly flammable, harmful, irritant
R Phrase 
R 11-20-37/38-41
Highly flammable.Harmful by inhalation.Irritating to respiratory system and skin.Risk of serious damage to eyes.
S Phrase 
S 16-26-39
Keep away from sources of ignition - No smoking.In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Wear eye/face protection.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?