EMAIL THIS PAGE TO A FRIEND
8.14657 EMD Millipore

Pinane

Pinane for synthesis. CAS 473-55-2, molar mass 138.25 g/mol., for synthesis

Synonym: 2,6,6-Trimethylbicyclo[3.1.1]heptane, Dihydropinene

  • CAS Number 473-55-2

  • Linear Formula C₁₀H₁₈

  • Molecular Weight 138.25 g/mol

  •  EC Number 207-467-1

Purchase

Properties

Related Categories Biochemicals and Reagents More...
bp   168 °C
expl. lim.     0.7 - 7.2 %(V)
vapor pressure   3.33 hPa (20 °C)
solubility   4.73 mg/l (calculated)
flash point   42 °C
mp   -53 °C
LD 50 oral   LD50 Rat > 5000 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.854 - 0.859
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2902 19 00
Hazard Statement(s) 
H226: Flammable liquid and vapour.
H411: Toxic to aquatic life with long lasting effects.
Precautionary Statement(s) 
P210: Keep away from heat/sparks/open flames/hot surfaces. No smoking.
P262: Do not get in eyes, on skin, or on clothing.
Storage class 
3 Flammable liquids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Pictogram(s) 
GHS09  GHS09
Hazard Symbols 
Dangerous for the environment  Dangerous for the environment
Declaration (railroad and road) ADR, RID 
UN 1993 , 3, III
Declaration (transport by air) IATA-DGR 
UN 1993 , 3, III
Declaration (transport by sea) IMDG-Code 
UN 1993 , 3, III
Categories of danger 
flammable, dangerous for the environment
R Phrase 
R 10-51/53
Flammable.Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
S Phrase 
S 16-61
Keep away from sources of ignition - No smoking.Avoid release to the environment. Refer to special instructions/ Safety data sheets.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?