• USA Home
  • 8.14660 - 4-tert-Butylbenzyl alcohol

EMAIL THIS PAGE TO A FRIEND
8.14660 EMD Millipore

4-tert-Butylbenzyl alcohol

4-tert-Butylbenzyl alcohol for synthesis. CAS 877-65-6, pH 7 (H₂O)., for synthesis

  • CAS Number 877-65-6

  • Linear Formula C₁₁H₁₆O

  • Molecular Weight 164.25 g/mol

  •  EC Number 212-894-1

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8146600025)
bp   140 °C (27 hPa)
expl. lim.     1.1 - 9.5 %(V)
vapor pressure   <1 hPa (20 °C)
density   1.05 g/cm3 (20 °C)
flash point   128 °C DIN 51758
autoignition temp.   405 °C DIN 51794
pH   7 (H₂O)
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.965 - 0.970
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2906 29 00
Hazard Statement(s) 
H315: Causes skin irritation.
H319: Causes serious eye irritation.
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Categories of danger 
irritant
R Phrase 
R 36/38
Irritating to eyes and skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?