EMAIL THIS PAGE TO A FRIEND
8.14661 EMD Millipore

2-Bromothiazole

2-Bromothiazole for synthesis. CAS 3034-53-5, molar mass 164.01 g/mol., for synthesis

  • CAS Number 3034-53-5

  • Linear Formula C₃H₂BrNS

  • Molecular Weight 164.01 g/mol

  •  EC Number 221-229-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 ml in Glass bottle (8146610005)
bp   70 - 71 °C (27 hPa)
density   1.84 g/cm3 (20 °C)
flash point   63 °C
shipped in   ambient
storage temp.   Store at +2°C to +8°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.845 - 1.850
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2934 10 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Declaration (railroad and road) ADR, RID 
Kein Gefahrgut
Declaration (transport by air) IATA-DGR 
No Dangerous Good
Declaration (transport by sea) IMDG-Code 
No Dangerous Good
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?