EMAIL THIS PAGE TO A FRIEND
8.14705 EMD Millipore

Acetylenediurea

Acetylenediurea for synthesis. CAS 496-46-8, molar mass 142.11 g/mol., for synthesis

Synonym: Glykuril

  • CAS Number 496-46-8

  • Linear Formula C₄H₆N₄O₂

  • Molecular Weight 142.11 g/mol

  •  EC Number 207-821-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   50 g in Plastic bottle (8147050050)
solubility   15 g/l
bulk density   800 kg/m3
LD 50 oral   LD50 Rat > 2000 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (ex N): ≥ 97.0 %
  Water (K. F.): ≤ 1.0 %
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 99 80
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?