• USA Home
  • 8.14706 - Methyl glycinate hydrochloride

EMAIL THIS PAGE TO A FRIEND
8.14706 EMD Millipore

Methyl glycinate hydrochloride

Methyl glycinate hydrochloride for synthesis. CAS 5680-79-5, EC Number 227-139-1, chemical formula NH₂CH₂CO₂CH₃*HCl., for synthesis

Synonym: Glycine methyl ester monohydrochloride

  • CAS Number 5680-79-5

  • Linear Formula C₃H₈ClNO₂

  • Molecular Weight 125.55 g/mol

  •  EC Number 227-139-1

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   500 g in Plastic bottle (8147060500)
mp   167 - 171 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (argentometric): ≥ 99.0 %
  Melting range (lower value): ≥ 167 °C
  Melting range (upper value): ≤ 171 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2922 49 85
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?