• USA Home
  • 8.14708 - p-tert-Butylcalix[6]arene

EMAIL THIS PAGE TO A FRIEND
8.14708 EMD Millipore

p-tert-Butylcalix[6]arene

p-tert-Butylcalix[6]arene for synthesis. CAS 78092-53-2, molar mass 973.38 g/mol., for synthesis

Synonym: 5,11,17,23,29,35-Hexa-tert-butyl-37,38,39,40,41,42-hexahydroxycalix[6]arene

  • CAS Number 78092-53-2

  • Linear Formula C₆₆H₈₄O₆

  • Molecular Weight 973.38 g/mol

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   1 g in Glass bottle (8147080001)
mp   380 - 381 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (to C, H): ≥ 98.0 %
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2907 29 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?