• USA Home
  • 8.14714 - 2-Fluorobenzaldehyde

EMAIL THIS PAGE TO A FRIEND
8.14714 EMD Millipore

2-Fluorobenzaldehyde

2-Fluorobenzaldehyde for synthesis. CAS 446-52-6, molar mass 124.11 g/mol., for synthesis

  • CAS Number 446-52-6

  • Linear Formula C₇H₅FO

  • Molecular Weight 124.11 g/mol

  •  EC Number 207-171-2

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 ml in Glass bottle (8147140010)
bp   172 - 174 °C (1013 hPa)
density   1.19 g/cm3 (20 °C)
flash point   55 °C
mp   -44.5 °C
LD 50 oral   LD50 Rat 1900 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.182 - 1.187
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2913 00 00
Hazard Statement(s) 
H226: Flammable liquid and vapour.
H302: Harmful if swallowed.
H315: Causes skin irritation.
Precautionary Statement(s) 
P210: Keep away from heat.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
Storage class 
3 Flammable liquids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Declaration (railroad and road) ADR, RID 
UN 1989 , 3, III
Declaration (transport by air) IATA-DGR 
UN 1989 , 3, III
Declaration (transport by sea) IMDG-Code 
UN 1989 , 3, III
Categories of danger 
flammable, harmful, irritant
R Phrase 
R 10-22-38
Flammable.Harmful if swallowed.Irritating to skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?