• USA Home
  • 8.14721 - 4-Acetamido-2,2,6,6-tetramethyl-N-piperidinyloxy

EMAIL THIS PAGE TO A FRIEND
8.14721 EMD Millipore

4-Acetamido-2,2,6,6-tetramethyl-N-piperidinyloxy

4-Acetamido-2,2,6,6-tetramethyl-N-piperidinyloxy (free radical) for synthesis. CAS 14691-89-5, molar mass 213.3 g/mol., (free radical) for synthesis

Synonym: 4-Acetamido-2,2,6,6-TEMPO

  • CAS Number 14691-89-5

  • Linear Formula C₁₁H₂₁N₂O₂

  • Molecular Weight 213.3 g/mol

  •  EC Number 423-840-3

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   1 g in Glass bottle (8147210001)
mp   141 - 145 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 137 °C
  Melting range (upper value): ≤ 143 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 39 99
Hazard Statement(s) 
H302: Harmful if swallowed.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Categories of danger 
harmful
R Phrase 
R 22
Harmful if swallowed.
S Phrase 
S 22
Do not breathe dust.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?