EMAIL THIS PAGE TO A FRIEND
8.14728 EMD Millipore

N-Cbz-Glycine

N-Cbz-Glycine for synthesis. CAS 1138-80-3, molar mass 209.2 g/mol., for synthesis

Synonym: N-Carbobenzyloxyglycine

  • CAS Number 1138-80-3

  • Linear Formula C₁₀H₁₁NO₄

  • Molecular Weight 209.2 g/mol

  •  EC Number 214-516-0

Purchase

Properties

Related Categories Amino Acids, Biochemicals and Reagents More...
packaging   25 g in Glass bottle (8147280025)
mp   118 - 122 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 97.0 % (a/a)
  Melting range (lower value): ≥ 116 °C
  Melting range (upper value): ≤ 119 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2924 29 70
RTECS 
MB9129000
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?