• USA Home
  • 8.14730 - N,N-Diethyl acetoacetamide

EMAIL THIS PAGE TO A FRIEND
8.14730 EMD Millipore

N,N-Diethyl acetoacetamide

N,N-Diethyl acetoacetamide for synthesis. CAS 2235-46-3, pH 5 - 7 (10 g/l, H₂O, 20 °C)., for synthesis

Synonym: Acetoacetic acid diethylamide

  • CAS Number 2235-46-3

  • Linear Formula C₈H₁₅NO₂

  • Molecular Weight 157.21 g/mol

  •  EC Number 218-792-3

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 ml in Glass bottle (8147300100)
bp   213 - 218 °C (1013 hPa)
expl. lim.     1 %(V)
vapor pressure   1.0 hPa (50 °C)
density   0.99 g/cm3 (20 °C)
flash point   101 °C DIN 51758
autoignition temp.   220 °C
mp   <-73 °C
pH   5 - 7 (10 g/l, H₂O, 20 °C)
LD 50 oral   LD50 Rat 4760 mg/kg
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.993 - 0.997
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2924 19 00
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?