• USA Home
  • 8.14736 - 2-Tetrahydrofurfurylamine

EMAIL THIS PAGE TO A FRIEND
8.14736 EMD Millipore

2-Tetrahydrofurfurylamine

2-Tetrahydrofurfurylamine for synthesis. CAS 4795-29-3, molar mass 101.15 g/mol., for synthesis

Synonym: 2-Aminomethyltetrahydrofuran

  • CAS Number 4795-29-3

  • Linear Formula C₅H₁₁NO

  • Molecular Weight 101.15 g/mol

  •  EC Number 225-351-9

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   500 ml in Glass bottle (8147360500)
bp   153 - 154 °C (1000 hPa)
expl. lim.     1.6 - 14.1 %(V)
density   0.98 g/cm3 (20 °C)
flash point   43 °C
autoignition temp.   270 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.975 - 0.979
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2932 19 00
Hazard Statement(s) 
H226: Flammable liquid and vapour.
H315: Causes skin irritation.
H319: Causes serious eye irritation.
Precautionary Statement(s) 
P210: Keep away from heat.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
3 Flammable liquids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Declaration (railroad and road) ADR, RID 
UN 2943 , 3, III
Declaration (transport by air) IATA-DGR 
UN 2943 , 3, III
Declaration (transport by sea) IMDG-Code 
UN 2943 , 3, III
Categories of danger 
flammable, irritant
R Phrase 
R 10-36/38
Flammable.Irritating to eyes and skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?