• USA Home
  • 8.14738 - 2-(2-Chloroethyl)-1-methylpiperidine hydrochloride

EMAIL THIS PAGE TO A FRIEND
8.14738 EMD Millipore

2-(2-Chloroethyl)-1-methylpiperidine hydrochloride

2-(2-Chloroethyl)-1-methylpiperidine hydrochloride for synthesis. CAS 58878-37-8, chemical formula C₈H₁₆ClN*HCl., for synthesis

  • CAS Number 58878-37-8

  • Linear Formula C₈H₁₇Cl₂N

  • Molecular Weight 198.14 g/mol

  •  EC Number 261-480-7

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 g in Plastic bottle (8147380010)
mp   133 - 137 °C
pH   5.9 (H₂O, 20 °C)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (argentometric): ≥ 98.0 %
  Melting range (lower value): ≥ 128 °C
  Melting range (upper value): ≤ 132 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 39 99
Hazard Statement(s) 
H302: Harmful if swallowed.
H315: Causes skin irritation.
H317: May cause an allergic skin reaction.
H319: Causes serious eye irritation.
H335: May cause respiratory irritation.
Precautionary Statement(s) 
P262: Do not get in eyes, on skin, or on clothing.
P280: Wear protective gloves.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P304 + P340: IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Categories of danger 
harmful, irritant, sensitizing
R Phrase 
R 22-36/37/38-43
Harmful if swallowed.Irritating to eyes, respiratory system and skin.May cause sensitisation by skin contact.
S Phrase 
S 24-37
Avoid contact with skin.Wear suitable gloves.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?