• USA Home
  • 8.14744 - 4-Fluorobenzyl alcohol

EMAIL THIS PAGE TO A FRIEND
8.14744 EMD Millipore

4-Fluorobenzyl alcohol

4-Fluorobenzyl alcohol for synthesis. CAS 459-56-3, molar mass 126.13 g/mol., for synthesis

  • CAS Number 459-56-3

  • Linear Formula C₇H₇FO

  • Molecular Weight 126.13 g/mol

  •  EC Number 207-292-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8147440025)
bp   79 - 81 °C (9 hPa)
density   1.156 g/cm3 (20 °C)
flash point   90 °C
mp   22 - 23 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.160 - 1.164
  Identity (IR): passes test
  Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2906 29 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?