• USA Home
  • 8.14751 - Decamethylcyclopentasiloxane

EMAIL THIS PAGE TO A FRIEND
8.14751 EMD Millipore

Decamethylcyclopentasiloxane

Decamethylcyclopentasiloxane for synthesis. CAS 541-02-6, molar mass 370.78 g/mol., for synthesis

  • CAS Number 541-02-6

  • Linear Formula C₁₀H₃₀O₅Si₅

  • Molecular Weight 370.78 g/mol

  •  EC Number 208-764-9

Purchase

Properties

expl. lim.     0.52 - 7 %(V)
vapor pressure   0.16 hPa (20 °C)
solubility   <0.0001 g/l (calculated)
density   0.96 g/cm3 (20 °C)
flash point   NULL °C DIN 51755 Part 1
autoignition temp.   NULL °C DIN 51794
mp   -36 °C
LD 50 oral   LD50 Rat > 24134 mg/kg
LD 50 dermal   LD50 Rabbit > 15200 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Refractive index (n 25°/D): 1.394 - 1.396
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2931 90 00
RTECS 
GY5945200
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?