EMAIL THIS PAGE TO A FRIEND
8.14756 EMD Millipore

Citrazinic acid

Citrazinic acid for synthesis. CAS 99-11-6, pH 3.1 - 3.5 (100 g/l, H₂O) (slurry)., for synthesis

  • CAS Number 99-11-6

  • Linear Formula C₆H₅NO₄

  • Molecular Weight 155.11 g/mol

  •  EC Number 202-731-2

Purchase

Properties

Related Categories Acids, Acids & Bases, Chemical Synthesis, Synthetic Reagents
packaging   100 g in Plastic bottle (8147560100)
density   0.65 g/cm3
mp   >330 °C (decomposition)
pH   3.1 - 3.5 (100 g/l, H₂O) (slurry)
LD 50 oral   LD50 Rat > 3200 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 98.0 % (a/a)
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 39 99
RTECS 
NS1400000
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?