EMAIL THIS PAGE TO A FRIEND
8.14772 EMD Millipore

Triethyl borate

Triethyl borate for synthesis. CAS 150-46-9, EC Number 205-760-9, chemical formula (CH₃CH₂O)₃B., for synthesis

  • CAS Number 150-46-9

  • Linear Formula C₆H₁₅BO₃

  • Molecular Weight 145.99 g/mol

  •  EC Number 205-760-9

Purchase

Properties

packaging   100 ml in Glass bottle (8147720100)
bp   117 - 118 °C (1013 hPa)
density   0.858 g/cm3
flash point   11 °C
mp   -84.8 °C
viscosity kinematic   0.503 mm2/s (40 °C)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.5 % (a/a)
  Density (d 20 °C/ 4 °C): 0.862 - 0.866
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2920 90 70
Hazard Statement(s) 
H225: Highly flammable liquid and vapour.
Precautionary Statement(s) 
P210: Keep away from heat/sparks/open flames/hot surfaces. No smoking.
P233: Keep container tightly closed.
P260: Do not breathe vapour.
RTECS 
ED5075000
Storage class 
3 Flammable liquids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Symbols 
Flammable  Flammable
Declaration (railroad and road) ADR, RID 
UN 1176 , 3, II
Declaration (transport by air) IATA-DGR 
UN 1176 , 3, II
Declaration (transport by sea) IMDG-Code 
UN 1176 , 3, II
Categories of danger 
highly flammable
R Phrase 
R 11
Highly flammable.
S Phrase 
S 7-16-23
Keep container tightly closed.Keep away from sources of ignition - No smoking.Do not breathe vapour.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?