• USA Home
  • 8.14775 - Boron trifluoride-acetic acid-complex

EMAIL THIS PAGE TO A FRIEND
8.14775 EMD Millipore

Boron trifluoride-acetic acid-complex

Boron trifluoride-acetic acid-complex for synthesis. CAS 373-61-5, molar mass 187.91 g/mol., for synthesis

  • CAS Number 373-61-5

  • Linear Formula C₄H₈O₄ * BF₃

  • Molecular Weight 187.91 g/mol

  •  EC Number 206-768-5

Purchase

Properties

bp   143 - 148 °C (1013 hPa)
vapor pressure   1 hPa (20 °C)
density   1.353 g/cm3 (20 °C)
flash point   83 °C
mp   -47 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (acidimetric): 33.0 - 37.0 %
  Density (d 20 °C/ 4 °C): 1.340 - 1.350

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2942 00 00
Hazard Statement(s) 
H314: Causes severe skin burns and eye damage.
H330: Fatal if inhaled.
Precautionary Statement(s) 
P280: Wear protective gloves/ protective clothing/ eye protection/ face protection.
P301 + P330 + P331: IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
P304 + P340: IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
Storage class 
6.1A Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK 
WGK 1 slightly water endangering
Disposal 
23
Hydrogen fluoride and inorganic fluorides as well as solutions thereof must be handled with extreme caution. Do not permit contact of any kind under any circumstances and be sure to work under an efficient fume cupboard with the front screen closed. Residues dissolved in water can be precipitated as calcium fluoride. Solids or precipitate: container I; filtrate: container D or E.
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Pictogram(s) 
GHS06  GHS06
Hazard Symbols 
Toxic  Toxic
Declaration (railroad and road) ADR, RID 
UN 1742 , 8, II
Declaration (transport by air) IATA-DGR 
UN 1742 , 8, II
Declaration (transport by sea) IMDG-Code 
UN 1742 , 8, II, Segregation Group: 1 (Acids)
Categories of danger 
toxic, corrosive
R Phrase 
R 23-34
Toxic by inhalation.Causes burns.
S Phrase 
S 26-36/37/39-45
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Wear suitable protective clothing, gloves and eye/face protection.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?