• USA Home
  • 8.14812 - ß-D-Glucose pentaacetate

EMAIL THIS PAGE TO A FRIEND
8.14812 EMD Millipore

ß-D-Glucose pentaacetate

ß-D-Glucose pentaacetate for synthesis. CAS 604-69-3, molar mass 390.34 g/mol., for synthesis

Synonym: Pentaacetyl-ß-D-glucose, 1,2,3,4,6-Penta-O-acetyl-ß-D-glucopyranose

  • CAS Number 604-69-3

  • Linear Formula C₁₆H₂₂O₁₁

  • Molecular Weight 390.34 g/mol

  •  EC Number 210-074-8

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 g in Glass bottle (8148120005)
solubility   <5 g/l
flash point   >150 °C
autoignition temp.   >500 °C
mp   130 - 132 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 130 °C
  Melting range (upper value): ≤ 132 °C
  Spec. rotation [α²0/D (c=10 in CHCl₃): +3.0 - +6.0 °
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2940 00 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?