• USA Home
  • 8.14817 - Isoquinoline-N-oxide

EMAIL THIS PAGE TO A FRIEND
8.14817 EMD Millipore

Isoquinoline-N-oxide

Isoquinoline-N-oxide for synthesis. CAS 1532-72-5, molar mass 145.16 g/mol., for synthesis

  • CAS Number 1532-72-5

  • Linear Formula C₉H₇NO

  • Molecular Weight 145.16 g/mol

  •  EC Number 216-242-7

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 g in Glass bottle (8148170005)
mp   102 - 105 °C (External MSDS)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 95.0 % (a/a)
  Identity (IR): passes test
  Melting point (after drying): 102 - 105 °C

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 49 90
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?