EMAIL THIS PAGE TO A FRIEND
8.14857 EMD Millipore

Methyl valerate

Methyl valerate for synthesis. CAS 624-24-8, molar mass 116.16 g/mol., for synthesis

Synonym: Methyl valerate, Valeric acid methyl ester, Pentanoic acid methyl ester

  • CAS Number 624-24-8

  • Linear Formula C₆H₁₂O₂

  • Molecular Weight 116.16 g/mol

  •  EC Number 210-838-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 ml in Glass bottle (8148570100)
bp   127 °C (1013 hPa)
vapor pressure   <=1 hPa
solubility   5 g/l
density   0.89 g/cm3 (20 °C)
flash point   29 °C
mp   -91 °C
LD 50 oral   LD50 Rat >= 5000 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.888 - 0.891
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2915 60 90
Hazard Statement(s) 
H226: Flammable liquid and vapour.
Precautionary Statement(s) 
P210: Keep away from heat.
Storage class 
3 Flammable liquids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Declaration (railroad and road) ADR, RID 
UN 3272 , 3, III
Declaration (transport by air) IATA-DGR 
UN 3272 , 3, III
Declaration (transport by sea) IMDG-Code 
UN 3272 , 3, III
Categories of danger 
flammable
R Phrase 
R 10
Flammable.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?