• USA Home
  • 8.14860 - 3-Methyl-4-nitrobenzoic acid

EMAIL THIS PAGE TO A FRIEND
8.14860 EMD Millipore

3-Methyl-4-nitrobenzoic acid

3-Methyl-4-nitrobenzoic acid for synthesis. CAS 3113-71-1, molar mass 181.15 g/mol., for synthesis

  • CAS Number 3113-71-1

  • Linear Formula C₈H₇NO₄

  • Molecular Weight 181.15 g/mol

  •  EC Number 221-479-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Glass bottle (8148600025)
mp   216 - 218 °C
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 215 °C
  Melting range (upper value): ≤ 218 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2916 39 90
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?