EMAIL THIS PAGE TO A FRIEND
8.14862 EMD Millipore

Diphenylglycin

Diphenylglycin for synthesis. CAS 3060-50-2, molar mass 227.26 g/mol., for synthesis

Synonym: Aminodiphenylacetic acid

  • CAS Number 3060-50-2

  • Linear Formula C₁₄H₁₃NO₂

  • Molecular Weight 227.26 g/mol

  •  EC Number 221-299-6

Purchase

Properties

Related Categories Amino Acids, Biochemicals and Reagents More...
packaging   5 g in Glass bottle (8148620005)
mp   244 - 247 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 235 °C
  Melting range (upper value): ≤ 240 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2922 49 85
Precautionary Statement(s) 
P260: Do not breathe dust.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
S Phrase 
S 22
Do not breathe dust.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?