EMAIL THIS PAGE TO A FRIEND
8.14880 EMD Millipore

1,9-Decadiene

1,9-Decadiene for synthesis. CAS 1647-16-1, molar mass 138.25 g/mol., for synthesis

  • CAS Number 1647-16-1

  • Linear Formula C₁₀H₁₈

  • Molecular Weight 138.25 g/mol

  •  EC Number 216-711-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 ml in Glass bottle (8148800005)
bp   169 °C (1013 hPa)
expl. lim.     0.7 %(V)
vapor pressure   5 hPa (20 °C)
density   0.75 g/cm3 (20 °C)
flash point   41 °C
autoignition temp.   250 °C
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.754 - 0.758
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2901 29 00
Hazard Statement(s) 
H226: Flammable liquid and vapour.
H410: Very toxic to aquatic life with long lasting effects.
Precautionary Statement(s) 
P210: Keep away from heat/sparks/open flames/hot surfaces. No smoking.
P262: Do not get in eyes, on skin, or on clothing.
P273: Avoid release to the environment.
Storage class 
3 Flammable liquids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Pictogram(s) 
GHS09  GHS09
Hazard Symbols 
Dangerous for the environment  Dangerous for the environment
Declaration (railroad and road) ADR, RID 
UN 3295 , 3, III
Declaration (transport by air) IATA-DGR 
UN 3295 , 3, III
Declaration (transport by sea) IMDG-Code 
UN 3295 , 3, III
Categories of danger 
flammable, dangerous for the environment
R Phrase 
R 10-50/53
Flammable.Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
S Phrase 
S 24/25-61
Avoid contact with skin and eyes.Avoid release to the environment. Refer to special instructions/ Safety data sheets.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?