• USA Home
  • 8.14881 - meso-1,2,3,4-Tetrabromobutane

EMAIL THIS PAGE TO A FRIEND
8.14881 EMD Millipore

meso-1,2,3,4-Tetrabromobutane

meso-1,2,3,4-Tetrabromobutane for synthesis. CAS 2657-67-2, molar mass 373.71 g/mol., for synthesis

  • CAS Number 2657-67-2

  • Linear Formula C₄H₆Br₄

  • Molecular Weight 373.71 g/mol

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Glass bottle (8148810025)
bp   181 °C (80 hPa)
vapor pressure   0.009 hPa (25 °C)
solubility   0.004 g/l
mp   116 - 119 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 116 °C
  Melting range (upper value): ≤ 119 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2903 39 19
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?