• USA Home
  • 8.14890 - 2-(Hydroxymethyl)-1,4-benzodioxane

EMAIL THIS PAGE TO A FRIEND
8.14890 EMD Millipore

2-(Hydroxymethyl)-1,4-benzodioxane

2-(Hydroxymethyl)-1,4-benzodioxane for synthesis. CAS 3663-82-9, molar mass 166.18 g/mol., for synthesis

Synonym: 1,4-Benzodioxan-2-methanol

  • CAS Number 3663-82-9

  • Linear Formula C₉H₁₀O₃

  • Molecular Weight 166.18 g/mol

  •  EC Number 222-921-9

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 g in Glass bottle (8148900005)
mp   90 °C
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Melting range (lower value): ≥ 87 °C
  Melting range (upper value): ≤ 90 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2932 99 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?