EMAIL THIS PAGE TO A FRIEND
8.14903 EMD Millipore

Butyl butyrate

Butyl butyrate for synthesis. CAS 109-21-7, molar mass 144.22 g/mol., for synthesis

Synonym: Butyric acid butyl ester, Butyl butanoate

  • CAS Number 109-21-7

  • Linear Formula C₈H₁₆O₂

  • Molecular Weight 144.22 g/mol

  •  EC Number 203-656-8

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 ml in Glass bottle (8149030100)
bp   164 - 165 °C (1013 hPa)
expl. lim.     1 %(V)
vapor pressure   4 hPa (20 °C)
solubility   0.50 g/l
density   0.87 g/cm3 (20 °C)
flash point   49 °C
autoignition temp.   454 °C
mp   -91 °C
LD 50 dermal   LD50 Rabbit > 5000 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2915 60 19
Hazard Statement(s) 
H226: Flammable liquid and vapour.
Precautionary Statement(s) 
P210: Keep away from heat.
RTECS 
ES8120000
Storage class 
3 Flammable liquids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Declaration (railroad and road) ADR, RID 
UN 1993 , 3, III
Declaration (transport by air) IATA-DGR 
UN 1993 , 3, III
Declaration (transport by sea) IMDG-Code 
UN 1993 , 3, III
Categories of danger 
flammable
R Phrase 
R 10
Flammable.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?