• USA Home
  • 8.14922 - 1-Bromo-2,4-difluorobenzene

EMAIL THIS PAGE TO A FRIEND
8.14922 EMD Millipore

1-Bromo-2,4-difluorobenzene

1-Bromo-2,4-difluorobenzene for synthesis. CAS 348-57-2, molar mass 192.99 g/mol., for synthesis

  • CAS Number 348-57-2

  • Linear Formula C₆H₃BrF₂

  • Molecular Weight 192.99 g/mol

  •  EC Number 206-479-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 ml in Glass bottle (8149220011)
density   1.704 - 1.708 g/cm3
flash point   51 °C
mp   -4 °C
refractive index   1.5050 - 1.5059 (20 °C, 589 nm)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.704 - 1.708
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2903 99 80
Hazard Statement(s) 
H226: Flammable liquid and vapour.
H315: Causes skin irritation.
H319: Causes serious eye irritation.
H335: May cause respiratory irritation.
Precautionary Statement(s) 
P210: Keep away from heat/sparks/open flames/hot surfaces. No smoking.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P304 + P340: IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
3 Flammable liquids
WGK 
WGK 2 water endangering
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Declaration (railroad and road) ADR, RID 
UN 2924 , 3 (8), II
Declaration (transport by air) IATA-DGR 
UN 2924 , 3 (8), II
Declaration (transport by sea) IMDG-Code 
UN 2924 , 3 (8), II
Categories of danger 
flammable, irritant
R Phrase 
R 10-36/37/38
Flammable.Irritating to eyes, respiratory system and skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?