• USA Home
  • 8.14928 - 5-(2-Hydroxyethyl)-4-methylthiazole

EMAIL THIS PAGE TO A FRIEND
8.14928 EMD Millipore

5-(2-Hydroxyethyl)-4-methylthiazole

5-(2-Hydroxyethyl)-4-methylthiazole for synthesis. CAS 137-00-8, molar mass 143.21 g/mol., for synthesis

Synonym: 2-(4-Methylthiazol-5-yl)ethanol, Sulfurol

  • CAS Number 137-00-8

  • Linear Formula C₆H₉NOS

  • Molecular Weight 143.21 g/mol

  •  EC Number 205-272-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8149280025)
bp   135 °C (9 hPa)
density   1.199 g/cm3 (20 °C)
flash point   112 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.197 - 1.201
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2934 10 00
Hazard Statement(s) 
H315: Causes skin irritation.
H319: Causes serious eye irritation.
H335: May cause respiratory irritation.
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P304 + P340: IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Categories of danger 
irritant
R Phrase 
R 36/37/38
Irritating to eyes, respiratory system and skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?