• USA Home
  • 8.14936 - 3-Fluorophenylboronic acid

EMAIL THIS PAGE TO A FRIEND
8.14936 EMD Millipore

3-Fluorophenylboronic acid

3-Fluorophenylboronic acid for synthesis. CAS 768-35-4, molar mass 139.92 g/mol., for synthesis

  • CAS Number 768-35-4

  • Linear Formula C₆H₆BFO₂

  • Molecular Weight 139.92 g/mol

Purchase

Properties

Related Categories Biochemicals and Reagents More...
mp   216 - 218 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 96.0 % (a/a)
  Melting range (lower value): ≥ 215 °C
  Melting range (upper value): ≤ 218 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2931 90 00
Hazard Statement(s) 
H314: Causes severe skin burns and eye damage.
H335: May cause respiratory irritation.
Precautionary Statement(s) 
P280: Wear protective gloves/ protective clothing/ eye protection/ face protection.
P301 + P330 + P331: IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
32
Chromatographic residues: Aggressive, explosive (e.g. peroxides) or toxic substances in chromatographic layers or sorbents must be eliminated using suitable methods (washing out, elution) prior to disposal. The solvents used for elution can in many cases be reutilized. Larger quantities of sorbents (e.g. from columns) should be freed from solvents (filtration, drying) and packed into rip-resistant plastic sachets; container I. TLC carrier materials and columns should be disposed of together with the corresponding waste materials (aluminium, glass, plastic); container K.
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Declaration (railroad and road) ADR, RID 
UN 3261 , 8, II
Declaration (transport by air) IATA-DGR 
UN 3261 , 8, II
Declaration (transport by sea) IMDG-Code 
UN 3261 , 8, II, Segregation Group: 1 (Acids)
Categories of danger 
irritant
R Phrase 
R 36/37/38
Irritating to eyes, respiratory system and skin.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?