• USA Home
  • 8.14938 - 3,4-Difluorophenylboronic acid

EMAIL THIS PAGE TO A FRIEND
8.14938 EMD Millipore

3,4-Difluorophenylboronic acid

3,4-Difluorophenylboronic acid for synthesis. CAS 168267-41-2, molar mass 157.91 g/mol., for synthesis

  • CAS Number 168267-41-2

  • Linear Formula C₆H₅BF₂O₂

  • Molecular Weight 157.91 g/mol

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Glass bottle (8149380025)
mp   271 - 279 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 96.0 % (a/a)
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2931 90 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?