• USA Home
  • 8.14957 - 4-Ethylphenylboronic acid

EMAIL THIS PAGE TO A FRIEND
8.14957 EMD Millipore

4-Ethylphenylboronic acid

4-Ethylphenylboronic acid for synthesis. CAS 63139-21-9, molar mass 149.99 g/mol., for synthesis

  • CAS Number 63139-21-9

  • Linear Formula C₈H₁₁BO₂

  • Molecular Weight 149.99 g/mol

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 g in Glass bottle (8149570005)
bp   507 - 511 °C (1013 hPa)
vapor pressure   <0.01 hPa (20 °C)
solubility   1.2 g/l
density   1.125 g/cm3 (20 °C)
autoignition temp.   >403 °C
mp   152.5 °C
LD 50 oral   LD50 Rat > 300 - 2000 mg/kg
LD 50 dermal   LD50 Rat > 2000 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 96.0 % (a/a)
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2931 90 00
Hazard Statement(s) 
H302: Harmful if swallowed.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Categories of danger 
harmful
R Phrase 
R 22
Harmful if swallowed.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?