• USA Home
  • 8.14960 - 4-(Trifluoromethoxy)benzaldehyde

EMAIL THIS PAGE TO A FRIEND
8.14960 EMD Millipore

4-(Trifluoromethoxy)benzaldehyde

4-(Trifluoromethoxy)benzaldehyde for synthesis. CAS 659-28-9, molar mass 190.12 g/mol., for synthesis

  • CAS Number 659-28-9

  • Linear Formula C₈H₅F₃O₂

  • Molecular Weight 190.12 g/mol

  •  EC Number 211-531-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 ml in Glass bottle (8149600010)
bp   185 °C
vapor pressure   3 hPa (20 °C)
density   1.33 g/cm3 (20 °C)
flash point   70.5 °C
autoignition temp.   220 °C DIN 51794
viscosity kinematic   1.7 mm2/s (20 °C)
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 96.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.330 - 1.335
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2913 00 00
Hazard Statement(s) 
H302: Harmful if swallowed.
H315: Causes skin irritation.
H319: Causes serious eye irritation.
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Categories of danger 
harmful, irritant
R Phrase 
R 22-36/38
Harmful if swallowed.Irritating to eyes and skin.
S Phrase 
S 26
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?