• USA Home
  • 8.14962 - 2-Fluoro-4-hydroxybenzonitrile

EMAIL THIS PAGE TO A FRIEND
8.14962 EMD Millipore

2-Fluoro-4-hydroxybenzonitrile

2-Fluoro-4-hydroxybenzonitrile for synthesis. CAS 82380-18-5, pH 4.2 (H₂O, 20 °C) (saturated aqueous solution)., for synthesis

Synonym: 4-Cyano-3-fluorophenol

  • CAS Number 82380-18-5

  • Linear Formula C₇H₄FNO

  • Molecular Weight 137.11 g/mol

  •  EC Number 422-810-7

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Glass bottle (8149620025)
solubility   4.7 g/l
density   1.45 g/cm3 (20 °C)
mp   124 °C
pH   4.2 (H₂O, 20 °C) (saturated aqueous solution)
bulk density   340 kg/m3
LD 50 oral   LD50 Rat 688 mg/kg
LD 50 dermal   LD50 Rat > 2000 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 122 °C
  Melting range (upper value): ≤ 125 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2926 90 70
Hazard Statement(s) 
H302: Harmful if swallowed.
H318: Causes serious eye damage.
H411: Toxic to aquatic life with long lasting effects.
Precautionary Statement(s) 
P273: Avoid release to the environment.
P280: Wear eye protection.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Pictogram(s) 
GHS09  GHS09
Hazard Symbols 
Dangerous for the environment  Dangerous for the environment
Hazard Symbols 
Harmful  Harmful
Declaration (railroad and road) ADR, RID 
UN 3077 , 9, III
Declaration (transport by air) IATA-DGR 
UN 3077 , 9, III
Declaration (transport by sea) IMDG-Code 
UN 3077 , 9, III, Marine Pollutant: P
Categories of danger 
harmful, irritant, dangerous for the environment
R Phrase 
R 22-41-51/53
Harmful if swallowed.Risk of serious damage to eyes.Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
S Phrase 
S 22-26-39-61
Do not breathe dust.In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Wear eye/face protection.Avoid release to the environment. Refer to special instructions/ Safety data sheets.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?